Assay Details
| Gene Symbol: QPCT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005630349.3 | XP_005630406.1 | 3 - 4 | 716 | 61 | |
| XM_005630350.2 | XP_005630407.1 | 4 - 5 | 636 | 61 | ||
| XM_022404634.1 | XP_022260342.1 | 5 - 6 | 1445 | 61 | ||
| XM_022404635.1 | XP_022260343.1 | 3 - 4 | 579 | 61 | ||
| XM_022404636.1 | XP_022260344.1 | 3 - 4 | 855 | 61 | ||
| GenBank mRNA | AF039440.1 | - | 2 - 3 | 331 | 61 | |
More Information
Set Membership: |
|

QPCT
Genomic Map