Assay Details
| Gene Symbol: NPY5R | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003118.1 | NP_001003118.1 | 1 | 1245 | 119 | |
| XM_014119227.2 | XP_013974702.1 | 2 | 2583 | 119 | ||
| XM_014119228.2 | XP_013974703.1 | 2 | 1348 | 119 | ||
| GenBank mRNA | AF049328.1 | - | 1 | 1245 | 119 | |
More Information
Set Membership: |
|

NPY5R
Genomic Map