Assay Details
| Gene Symbol: CHRM3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005618760.2 | XP_005618817.1 | 1805 | 66 | ||
| XM_005618761.2 | XP_005618818.1 | 1809 | 66 | |||
| XM_005618763.2 | XP_005618820.1 | 1742 | 66 | |||
| XM_005618766.2 | XP_005618823.1 | 1908 | 66 | |||
| XM_005618768.2 | XP_005618825.1 | 1834 | 66 | |||
| XM_005618769.2 | XP_005618826.1 | 1972 | 66 | |||
| XM_014112712.2 | XP_013968187.1 | 1746 | 66 | |||
| XM_014112713.2 | XP_013968188.1 | 2097 | 66 | |||
| XM_014112714.2 | XP_013968189.1 | 1633 | 66 | |||
| XM_022417355.1 | XP_022273063.1 | 2015 | 66 | |||
| XM_022417356.1 | XP_022273064.1 | 2075 | 66 | |||
| XM_022417357.1 | XP_022273065.1 | 2104 | 66 | |||
| XM_022417358.1 | XP_022273066.1 | 2170 | 66 | |||
| XM_022417359.1 | XP_022273067.1 | 2061 | 66 | |||
| XM_022417360.1 | XP_022273068.1 | 1898 | 66 | |||
| XM_022417361.1 | XP_022273069.1 | 1696 | 66 | |||
| XM_022417362.1 | XP_022273070.1 | 1843 | 66 | |||
| XM_022417363.1 | XP_022273071.1 | 1826 | 66 | |||
| XM_022417364.1 | XP_022273072.1 | 1440 | 66 | |||
| XM_546069.6 | XP_546069.4 | 1859 | 66 | |||
| GenBank mRNA | AF056305.1 | - | 260 | 66 | ||
More Information
Set Membership: |
|

CHRM3
Genomic Map