Assay Details
| Gene Symbol: FLT3LG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003350.1 | NP_001003350.1 | 4 - 5 | 375 | 67 | |
| XM_005615738.3 | XP_005615795.1 | 5 - 6 | 559 | 67 | ||
| XM_005615739.3 | XP_005615796.1 | 5 - 6 | 502 | 67 | ||
| XM_014114041.2 | XP_013969516.1 | 5 - 6 | 524 | 67 | ||
| XM_022417411.1 | XP_022273119.1 | 5 - 6 | 496 | 67 | ||
| XM_022417456.1 | XP_022273164.1 | 5 - 6 | 449 | 67 | ||
| GenBank mRNA | AF155148.1 | - | 4 - 5 | 375 | 67 | |
More Information
Set Membership: |
|

FLT3LG
Genomic Map