Assay Details
| Gene Symbol: FLT3LG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003350.1 | NP_001003350.1 | 6 - 7 | 701 | 105 | |
| XM_005615738.3 | XP_005615795.1 | 7 - 8 | 885 | 105 | ||
| XM_005615739.3 | XP_005615796.1 | 7 - 8 | 828 | 105 | ||
| XM_014114041.2 | XP_013969516.1 | 7 - 8 | 850 | 105 | ||
| XM_022417411.1 | XP_022273119.1 | 7 - 8 | 822 | 105 | ||
| XM_022417428.1 | XP_022273136.1 | 7 - 8 | 797 | 105 | ||
| XM_022417456.1 | XP_022273164.1 | 7 - 8 | 775 | 105 | ||
| GenBank mRNA | AF155148.1 | - | 6 - 7 | 701 | 105 | |
More Information
Set Membership: |
|

FLT3LG
Genomic Map