Assay Details
| Gene Symbol: PTGER4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003054.1 | NP_001003054.1 | 1 - 2 | 231 | 111 | |
| GenBank mRNA | AF177934.1 | - | 1 - 2 | 231 | 111 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001003054.1 transcripts.
Set Membership: |
|

PTGER4
Genomic Map