Assay Details
| Gene Symbol: PTGER4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003054.1 | NP_001003054.1 | 2 - 3 | 1168 | 68 | |
| GenBank mRNA | AF177934.1 | - | 2 - 3 | 1168 | 68 | |
More Information
Set Membership: |
|

PTGER4
Genomic Map