Assay Details
| Gene Symbol: FGFR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003336.1 | NP_001003336.1 | 11 - 12 | 1792 | 101 | |
| XM_005637445.3 | XP_005637502.1 | 15 - 16 | 2484 | 101 | ||
| XM_005637446.3 | XP_005637503.1 | 15 - 16 | 2477 | 101 | ||
| XM_005637447.3 | XP_005637504.1 | 15 - 16 | 2479 | 101 | ||
| XM_005637448.3 | XP_005637505.1 | 15 - 16 | 2472 | 101 | ||
| XM_005637449.3 | XP_005637506.1 | 14 - 15 | 2405 | 101 | ||
| XM_005637450.3 | XP_005637507.1 | 14 - 15 | 2401 | 101 | ||
| XM_005637451.3 | XP_005637508.1 | 14 - 15 | 2394 | 101 | ||
| XM_005637452.3 | XP_005637509.1 | 15 - 16 | 2488 | 101 | ||
| XM_005637453.3 | XP_005637510.1 | 14 - 15 | 2399 | 101 | ||
| XM_005637454.3 | XP_005637511.1 | 12 - 13 | 2057 | 101 | ||
| XM_005637456.3 | XP_005637513.1 | 12 - 13 | 2046 | 101 | ||
| XM_014108754.2 | XP_013964229.1 | 14 - 15 | 2399 | 101 | ||
| XM_014108755.2 | XP_013964230.1 | 13 - 14 | 2136 | 101 | ||
| XM_014108756.2 | XP_013964231.1 | 13 - 14 | 2130 | 101 | ||
| XM_014108757.2 | XP_013964232.1 | 12 - 13 | 2051 | 101 | ||
| GenBank mRNA | AF211257.1 | - | 11 - 12 | 1792 | 101 | |
More Information
Set Membership: |
|

FGFR2
Genomic Map