Assay Details
| Gene Symbol: FGFR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003336.1 | NP_001003336.1 | 1 - 2 | 260 | 86 | |
| XM_005637454.3 | XP_005637511.1 | 2 - 3 | 525 | 86 | ||
| XM_005637456.3 | XP_005637513.1 | 2 - 3 | 523 | 86 | ||
| XM_014108757.2 | XP_013964232.1 | 2 - 3 | 522 | 86 | ||
| GenBank mRNA | AF211257.1 | - | 1 - 2 | 260 | 86 | |
More Information
Set Membership: |
|

FGFR2
Genomic Map