Assay Details
| Gene Symbol: FGFR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003336.1 | NP_001003336.1 | 2 - 3 | 425 | 72 | |
| XM_005637445.3 | XP_005637502.1 | 6 - 7 | 1117 | 72 | ||
| XM_005637446.3 | XP_005637503.1 | 6 - 7 | 1113 | 72 | ||
| XM_005637447.3 | XP_005637504.1 | 6 - 7 | 1118 | 72 | ||
| XM_005637448.3 | XP_005637505.1 | 6 - 7 | 1114 | 72 | ||
| XM_005637449.3 | XP_005637506.1 | 5 - 6 | 1038 | 72 | ||
| XM_005637450.3 | XP_005637507.1 | 5 - 6 | 1040 | 72 | ||
| XM_005637451.3 | XP_005637508.1 | 5 - 6 | 1036 | 72 | ||
| XM_005637452.3 | XP_005637509.1 | 6 - 7 | 1121 | 72 | ||
| XM_005637453.3 | XP_005637510.1 | 5 - 6 | 1041 | 72 | ||
| XM_005637454.3 | XP_005637511.1 | 3 - 4 | 690 | 72 | ||
| XM_005637456.3 | XP_005637513.1 | 3 - 4 | 688 | 72 | ||
| XM_014108754.2 | XP_013964229.1 | 5 - 6 | 1035 | 72 | ||
| XM_014108755.2 | XP_013964230.1 | 4 - 5 | 769 | 72 | ||
| XM_014108756.2 | XP_013964231.1 | 4 - 5 | 766 | 72 | ||
| XM_014108757.2 | XP_013964232.1 | 3 - 4 | 687 | 72 | ||
| GenBank mRNA | AF211257.1 | - | 2 - 3 | 425 | 72 | |
More Information
Set Membership: |
|

FGFR2
Genomic Map