Assay Details
| Gene Symbol: FGFR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003336.1 | NP_001003336.1 | 3 - 4 | 557 | 75 | |
| XM_005637445.3 | XP_005637502.1 | 7 - 8 | 1249 | 75 | ||
| XM_005637446.3 | XP_005637503.1 | 7 - 8 | 1245 | 75 | ||
| XM_005637447.3 | XP_005637504.1 | 7 - 8 | 1250 | 75 | ||
| XM_005637448.3 | XP_005637505.1 | 7 - 8 | 1246 | 75 | ||
| XM_005637449.3 | XP_005637506.1 | 6 - 7 | 1170 | 75 | ||
| XM_005637450.3 | XP_005637507.1 | 6 - 7 | 1172 | 75 | ||
| XM_005637451.3 | XP_005637508.1 | 6 - 7 | 1168 | 75 | ||
| XM_005637452.3 | XP_005637509.1 | 7 - 8 | 1253 | 75 | ||
| XM_005637453.3 | XP_005637510.1 | 6 - 7 | 1173 | 75 | ||
| XM_005637454.3 | XP_005637511.1 | 4 - 5 | 822 | 75 | ||
| XM_005637456.3 | XP_005637513.1 | 4 - 5 | 820 | 75 | ||
| XM_014108754.2 | XP_013964229.1 | 6 - 7 | 1167 | 75 | ||
| XM_014108755.2 | XP_013964230.1 | 5 - 6 | 901 | 75 | ||
| XM_014108756.2 | XP_013964231.1 | 5 - 6 | 898 | 75 | ||
| XM_014108757.2 | XP_013964232.1 | 4 - 5 | 819 | 75 | ||
| GenBank mRNA | AF211257.1 | - | 3 - 4 | 557 | 75 | |
More Information
Set Membership: |
|

FGFR2
Genomic Map