Assay Details
| Gene Symbol: FGFR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003336.1 | NP_001003336.1 | 4 - 5 | 742 | 66 | |
| XM_005637445.3 | XP_005637502.1 | 8 - 9 | 1434 | 66 | ||
| XM_005637447.3 | XP_005637504.1 | 8 - 9 | 1435 | 66 | ||
| XM_005637449.3 | XP_005637506.1 | 7 - 8 | 1355 | 66 | ||
| XM_005637450.3 | XP_005637507.1 | 7 - 8 | 1357 | 66 | ||
| XM_005637452.3 | XP_005637509.1 | 8 - 9 | 1438 | 66 | ||
| XM_005637454.3 | XP_005637511.1 | 5 - 6 | 1007 | 66 | ||
| XM_014108755.2 | XP_013964230.1 | 6 - 7 | 1086 | 66 | ||
| GenBank mRNA | AF211257.1 | - | 4 - 5 | 742 | 66 | |
More Information
Set Membership: |
|

FGFR2
Genomic Map