Assay Details
| Gene Symbol: OCLN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003195.1 | NP_001003195.1 | 7 - 8 | 1541 | 92 | |
| GenBank mRNA | AF246976.1 | - | 8 - 9 | 1663 | 92 | |
| U49221.1 | - | 7 - 8 | 1541 | 92 | ||
More Information
Set Membership: |
|

OCLN
Genomic Map