Assay Details
| Gene Symbol: FGFR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005637445.3 | XP_005637502.1 | 4 - 5 | 871 | 71 | |
| XM_005637446.3 | XP_005637503.1 | 4 - 5 | 867 | 71 | ||
| XM_005637447.3 | XP_005637504.1 | 4 - 5 | 872 | 71 | ||
| XM_005637448.3 | XP_005637505.1 | 4 - 5 | 868 | 71 | ||
| XM_005637449.3 | XP_005637506.1 | 3 - 4 | 792 | 71 | ||
| XM_005637450.3 | XP_005637507.1 | 3 - 4 | 794 | 71 | ||
| XM_005637451.3 | XP_005637508.1 | 3 - 4 | 790 | 71 | ||
| XM_005637452.3 | XP_005637509.1 | 4 - 5 | 875 | 71 | ||
| XM_005637453.3 | XP_005637510.1 | 3 - 4 | 795 | 71 | ||
| XM_014108754.2 | XP_013964229.1 | 3 - 4 | 789 | 71 | ||
| GenBank mRNA | AF345933.1 | - | 1 - 2 | 212 | 71 | |
More Information
Set Membership: |
|

FGFR2
Genomic Map