Assay Details
| Gene Symbol: FGFR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005637445.3 | XP_005637502.1 | 5 - 6 | 949 | 74 | |
| XM_005637446.3 | XP_005637503.1 | 5 - 6 | 945 | 74 | ||
| XM_005637447.3 | XP_005637504.1 | 5 - 6 | 950 | 74 | ||
| XM_005637448.3 | XP_005637505.1 | 5 - 6 | 946 | 74 | ||
| XM_005637449.3 | XP_005637506.1 | 4 - 5 | 870 | 74 | ||
| XM_005637450.3 | XP_005637507.1 | 4 - 5 | 872 | 74 | ||
| XM_005637451.3 | XP_005637508.1 | 4 - 5 | 868 | 74 | ||
| XM_005637452.3 | XP_005637509.1 | 5 - 6 | 953 | 74 | ||
| XM_005637453.3 | XP_005637510.1 | 4 - 5 | 873 | 74 | ||
| XM_014108754.2 | XP_013964229.1 | 4 - 5 | 867 | 74 | ||
| XM_014108755.2 | XP_013964230.1 | 3 - 4 | 601 | 74 | ||
| XM_014108756.2 | XP_013964231.1 | 3 - 4 | 598 | 74 | ||
| GenBank mRNA | AF345933.1 | - | 2 - 3 | 290 | 74 | |
More Information
Set Membership: |
|

FGFR2
Genomic Map