Assay Details
| Gene Symbol: LIF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001197073.1 | NP_001184002.1 | 2 - 3 | 354 | 73 | |
| XM_005636000.2 | XP_005636057.1 | 3 - 4 | 2396 | 73 | ||
| XM_022410147.1 | XP_022265855.1 | 4 - 5 | 1312 | 73 | ||
| GenBank mRNA | AF479881.1 | - | 1 - 2 | 48 | 73 | |
| AF512028.1 | - | 1 - 2 | 138 | 73 | ||
More Information
Set Membership: |
|

LIF
Genomic Map