Assay Details
| Gene Symbol: CDKN1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002957.2 | NP_001002957.2 | 1 - 2 | 476 | 142 | |
| XM_005636668.3 | XP_005636725.1 | 1 - 2 | 1128 | 142 | ||
| GenBank mRNA | AJ810861.1 | - | 1 - 2 | 476 | 142 | |
| AY455798.1 | - | 1 - 2 | 476 | 142 | ||
More Information
Set Membership: |
|

CDKN1B
Genomic Map