Assay Details
| Gene Symbol: CLN8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001012343.1 | NP_001012343.1 | 1 - 2 | 59 | 84 | |
| GenBank mRNA | AJ875419.1 | - | 1 - 2 | 59 | 84 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AJ875419.1, NM_001012343.1 transcripts.
Set Membership: |
|

CLN8
Genomic Map