Assay Details
| Gene Symbol: CLN8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001012343.1 | NP_001012343.1 | 2 - 3 | 733 | 70 | |
| XM_005640436.3 | XP_005640493.1 | 2 - 3 | 952 | 70 | ||
| XM_022414959.1 | XP_022270667.1 | 4 - 5 | 958 | 70 | ||
| XM_022414960.1 | XP_022270668.1 | 2 - 3 | 934 | 70 | ||
| GenBank mRNA | AJ875419.1 | - | 2 - 3 | 733 | 70 | |
More Information
Set Membership: |
|

CLN8
Genomic Map