Assay Details
| Gene Symbol: SSTR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001031816.1 | NP_001026986.1 | 1 | 718 | 76 | |
| XM_005623172.3 | XP_005623229.1 | 2 | 960 | 76 | ||
| XM_022421736.1 | XP_022277444.1 | 1 | 1062 | 76 | ||
| XM_022421737.1 | XP_022277445.1 | 1 | 1062 | 76 | ||
| GenBank mRNA | AY305400.1 | - | 1 | 552 | 76 | |
More Information
Set Membership: |
|

SSTR1
Genomic Map