Assay Details
| Gene Symbol: DEFB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113713.1 | NP_001107185.1 | 1 - 2 | 210 | 64 | |
| GenBank mRNA | DQ011970.1 | - | 1 - 2 | 210 | 64 | |
More Information
Set Membership: |
|

DEFB1
Genomic Map