Assay Details
| Gene Symbol: ACKR3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005635904.3 | XP_005635961.1 | 1 - 2 | 106 | 84 | |
| GenBank mRNA | X14048.1 | - | 64 | 84 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the X14048.1, XM_005635904.3 transcripts.
Set Membership: |
|

ACKR3
Genomic Map