Assay Details
| Gene Symbol: SULT1A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003223.1 | NP_001003223.1 | 3 - 4 | 346 | 72 | |
| XM_014114387.1 | XP_013969862.1 | 3 - 4 | 627 | 72 | ||
| XM_022419477.1 | XP_022275185.1 | 3 - 4 | 557 | 72 | ||
| GenBank mRNA | AY069922.1 | - | 3 - 4 | 322 | 72 | |
| D29807.1 | - | 3 - 4 | 346 | 72 | ||
More Information
Set Membership: |
|

SULT1A1
Genomic Map