Assay Details
| Gene Symbol: EPO | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006646.1 | NP_001006647.1 | 193 | 80 | ||
| GenBank mRNA | AB716701.1 | - | 151 | 80 | ||
| AY572971.1 | - | 193 | 80 | |||
| L13027.1 | - | 139 | 80 | |||
More Information
Set Membership: |
|

EPO
Genomic Map