Assay Details
| Gene Symbol: AKR1C3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001012344.1 | NP_001012344.1 | 231 | 71 | ||
| XM_005617078.3 | XP_005617135.1 | 554 | 71 | |||
| XM_005617079.3 | XP_005617136.1 | 397 | 71 | |||
| GenBank mRNA | AY875970.1 | - | 231 | 71 | ||
More Information
Set Membership: |
|

AKR1C3
Genomic Map