Assay Details
| Gene Symbol: IL24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022421680.1 | XP_022277388.1 | 466 | 62 | ||
| GenBank mRNA | DQ195089.1 | - | 318 | 62 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: IL24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022421680.1 | XP_022277388.1 | 466 | 62 | ||
| GenBank mRNA | DQ195089.1 | - | 318 | 62 | ||
Set Membership: |
|