Assay Details
| Gene Symbol: F7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001048033.1 | NP_001041498.1 | 6 - 7 | 625 | 67 | |
| XM_005634093.3 | XP_005634150.1 | 6 - 7 | 710 | 67 | ||
| XM_005634094.3 | XP_005634151.1 | 4 - 5 | 491 | 67 | ||
| GenBank mRNA | DQ223901.1 | - | 6 - 7 | 625 | 67 | |
More Information
Set Membership: |
|

F7
Genomic Map