Assay Details
| Gene Symbol: ADGRE3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001038666.1 | NP_001033755.1 | 1 - 2 | 99 | 92 | |
| XM_022406246.1 | XP_022261954.1 | 1 - 2 | 132 | 92 | ||
| GenBank mRNA | DQ227277.1 | - | 1 - 2 | 99 | 92 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001038666.1, XM_022406246.1 transcripts.
Set Membership: |
|

ADGRE3
Genomic Map