Assay Details
| Gene Symbol: RAB2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003318.2 | NP_001003318.1 | 7 - 8 | 550 | 129 | |
| XM_005637931.3 | XP_005637988.1 | 7 - 8 | 625 | 129 | ||
| XM_014109167.2 | XP_013964642.1 | 7 - 8 | 538 | 129 | ||
| GenBank mRNA | M35521.1 | - | 7 - 8 | 550 | 129 | |
More Information
Set Membership: |
|

RAB2A
Genomic Map