Assay Details
| Gene Symbol: GP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003371.2 | NP_001003371.2 | 2 - 3 | 234 | 63 | |
| GenBank mRNA | M64083.1 | - | 1 - 2 | 71 | 63 | |
More Information
Set Membership: |
|

GP2
Genomic Map