Assay Details
| Gene Symbol: CSF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003245.1 | NP_001003245.1 | 1 - 2 | 165 | 86 | |
| XM_005626371.2 | XP_005626428.1 | 2 - 3 | 1570 | 86 | ||
| XM_005626372.2 | XP_005626429.1 | 2 - 3 | 1465 | 86 | ||
| XM_022424894.1 | XP_022280602.1 | 2 - 3 | 1588 | 86 | ||
| XM_022424895.1 | XP_022280603.1 | 3 - 4 | 1103 | 86 | ||
| XM_022424896.1 | XP_022280604.1 | 3 - 4 | 1085 | 86 | ||
| GenBank mRNA | S49738.1 | - | 1 - 2 | 165 | 86 | |
More Information
Set Membership: |
|

CSF2
Genomic Map