Assay Details
| Gene Symbol: CSF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003245.1 | NP_001003245.1 | 2 - 3 | 204 | 85 | |
| XM_005626371.2 | XP_005626428.1 | 3 - 4 | 1609 | 85 | ||
| XM_005626372.2 | XP_005626429.1 | 3 - 4 | 1504 | 85 | ||
| XM_022424894.1 | XP_022280602.1 | 3 - 4 | 1627 | 85 | ||
| XM_022424895.1 | XP_022280603.1 | 4 - 5 | 1142 | 85 | ||
| XM_022424896.1 | XP_022280604.1 | 4 - 5 | 1124 | 85 | ||
| GenBank mRNA | S49738.1 | - | 2 - 3 | 204 | 85 | |
More Information
Set Membership: |
|

CSF2
Genomic Map