Assay Details
| Gene Symbol: TFPI | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003243.2 | NP_001003243.1 | 4 - 5 | 397 | 72 | |
| XM_005640183.3 | XP_005640240.1 | 6 - 7 | 910 | 72 | ||
| XM_022414647.1 | XP_022270355.1 | 6 - 7 | 763 | 72 | ||
| XM_022414648.1 | XP_022270356.1 | 5 - 6 | 632 | 72 | ||
| XM_022414649.1 | XP_022270357.1 | 4 - 5 | 511 | 72 | ||
| GenBank mRNA | S75369.1 | - | 4 - 5 | 398 | 72 | |
More Information
Set Membership: |
|

TFPI
Genomic Map