Assay Details
| Gene Symbol: RAC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003274.2 | NP_001003274.1 | 2 | 290 | 101 | |
| XM_005621021.1 | XP_005621078.1 | 1 | 350 | 101 | ||
| XM_022419478.1 | XP_022275186.1 | 1 | 341 | 101 | ||
| GenBank mRNA | X56390.1 | - | 290 | 101 | ||
More Information
Set Membership: |
|

RAC1
Genomic Map