Assay Details
| Gene Symbol: APOH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003265.2 | NP_001003265.1 | 999 | 67 | ||
| GenBank mRNA | X72933.1 | - | 999 | 67 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: APOH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003265.2 | NP_001003265.1 | 999 | 67 | ||
| GenBank mRNA | X72933.1 | - | 999 | 67 | ||
Set Membership: |
|