Assay Details
| Gene Symbol: GNB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003236.1 | NP_001003236.1 | 1 | 786 | 66 | |
| XM_005620339.2 | XP_005620396.1 | 1 | 1087 | 66 | ||
| XM_005620340.3 | XP_005620397.1 | 1 | 1126 | 66 | ||
| XM_005620341.3 | XP_005620398.1 | 1 | 1023 | 66 | ||
| GenBank mRNA | Z75134.1 | - | 1 | 786 | 66 | |
More Information
Set Membership: |
|

GNB1
Genomic Map