Assay Details
| Gene Symbol: LOC609288 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022405185.1 | XP_022260893.1 | 431 | 116 | ||
| GenBank mRNA | AB117154.1 | - | 58 | 116 | ||
| AB117159.1 | - | 58 | 116 | |||
| AB117160.1 | - | 58 | 116 | |||
| AB117161.1 | - | 58 | 116 | |||
| FR828700.1 | - | 254 | 116 | |||
More Information
Set Membership: |
|

LOC609288
Genomic Map