Assay Details
| Gene Symbol: CYP17A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005637679.2 | XP_005637736.1 | 1 - 2 | 366 | 79 | |
| XM_535000.5 | XP_535000.3 | 1 - 2 | 296 | 79 | ||
More Information
Set Membership: |
|

CYP17A1
Genomic Map