Assay Details
| Gene Symbol: AIP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014121247.2 | XP_013976722.1 | 4 - 5 | 842 | 112 | |
| XM_846748.5 | XP_851841.1 | 4 - 5 | 838 | 112 | ||
More Information
Set Membership: |
|

AIP
Genomic Map