Assay Details
| Gene Symbol: SEPTIN6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005641741.3 | XP_005641798.1 | 5 - 6 | 980 | 99 | |
| XM_014111689.2 | XP_013967164.1 | 5 - 6 | 980 | 99 | ||
| XM_014111690.1 | XP_013967165.1 | 5 - 6 | 979 | 99 | ||
| XM_014111691.2 | XP_013967166.1 | 5 - 6 | 980 | 99 | ||
More Information
Set Membership: |
|

SEPTIN6
Genomic Map