Assay Details
| Gene Symbol: LACC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005633914.3 | XP_005633971.1 | 2 - 3 | 896 | 90 | |
| XM_005633915.3 | XP_005633972.1 | 3 - 4 | 852 | 90 | ||
| XM_005633916.3 | XP_005633973.1 | 3 - 4 | 924 | 90 | ||
| XM_022408322.1 | XP_022264030.1 | 3 - 4 | 850 | 90 | ||
| XM_022408323.1 | XP_022264031.1 | 3 - 4 | 858 | 90 | ||
| XM_022408324.1 | XP_022264032.1 | 3 - 4 | 858 | 90 | ||
| XM_022408325.1 | XP_022264033.1 | 3 - 4 | 854 | 90 | ||
| XM_534129.4 | XP_534129.1 | 3 - 4 | 848 | 90 | ||
More Information
Set Membership: |
|

LACC1
Genomic Map