Assay Details
| Gene Symbol: NDUFA11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_533939.6 | XP_533939.2 | 2 - 3 | 272 | 59 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

NDUFA11| Gene Symbol: NDUFA11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_533939.6 | XP_533939.2 | 2 - 3 | 272 | 59 | |
Set Membership: |
|