Assay Details
| Gene Symbol: MRPL48 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022407700.1 | XP_022263408.1 | 7 - 8 | 462 | 73 | |
| XM_022407701.1 | XP_022263409.1 | 7 - 8 | 585 | 73 | ||
| XM_022407702.1 | XP_022263410.1 | 6 - 7 | 432 | 73 | ||
| XM_022407703.1 | XP_022263411.1 | 6 - 7 | 555 | 73 | ||
| XM_022407704.1 | XP_022263412.1 | 6 - 7 | 420 | 73 | ||
| XM_022407705.1 | XP_022263413.1 | 5 - 6 | 390 | 73 | ||
| XM_534016.6 | XP_534016.2 | 5 - 6 | 513 | 73 | ||
More Information
Set Membership: |
|

MRPL48
Genomic Map