Assay Details
| Gene Symbol: BBOX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022407945.1 | XP_022263653.1 | 6 - 7 | 982 | 98 | |
| XM_534096.6 | XP_534096.2 | 7 - 8 | 1174 | 98 | ||
More Information
Set Membership: |
|

BBOX1
Genomic Map