Assay Details
| Gene Symbol: LOC476718 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022407379.1 | XP_022263087.1 | 45 - 46 | 15951 | 92 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC476718| Gene Symbol: LOC476718 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022407379.1 | XP_022263087.1 | 45 - 46 | 15951 | 92 | |
Set Membership: |
|