Assay Details
| Gene Symbol: WDR83OS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_533908.6 | XP_533908.1 | 3 - 4 | 342 | 83 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

WDR83OS| Gene Symbol: WDR83OS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_533908.6 | XP_533908.1 | 3 - 4 | 342 | 83 | |
Set Membership: |
|