Assay Details
| Gene Symbol: SELENOT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001164487.1 | NP_001157959.1 | 1 - 2 | 230 | 56 | |
| GenBank mRNA | DN397021.1 | - | 1 - 2 | 263 | 56 | |
| DN425872.1 | - | 1 - 2 | 188 | 56 | ||
More Information
Set Membership: |
|

SELENOT
Genomic Map