Assay Details
| Gene Symbol: TCEA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005637986.3 | XP_005638043.1 | 7 - 8 | 854 | 71 | |
| XM_005637987.3 | XP_005638044.1 | 7 - 8 | 852 | 71 | ||
| XM_005637989.3 | XP_005638046.1 | 7 - 8 | 1247 | 71 | ||
More Information
Set Membership: |
|

TCEA1
Genomic Map