Assay Details
| Gene Symbol: PRICKLE1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003639965.4 | XP_003640013.1 | 6 - 7 | 878 | 73 | |
| XM_005636965.3 | XP_005637022.1 | 7 - 8 | 1101 | 73 | ||
| XM_014108553.2 | XP_013964028.1 | 6 - 7 | 1040 | 73 | ||
More Information
Set Membership: |
|

PRICKLE1
Genomic Map