Assay Details
| Gene Symbol: PUS7L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005636961.3 | XP_005637018.2 | 8 - 9 | 3014 | 78 | |
| XM_005636962.3 | XP_005637019.2 | 7 - 8 | 1002 | 78 | ||
| XM_022411046.1 | XP_022266754.1 | 9 - 10 | 2916 | 78 | ||
| XM_022411047.1 | XP_022266755.1 | 9 - 10 | 2706 | 78 | ||
| XM_022411048.1 | XP_022266756.1 | 8 - 9 | 2536 | 78 | ||
| XM_022411049.1 | XP_022266757.1 | 8 - 9 | 1812 | 78 | ||
| XM_022411050.1 | XP_022266758.1 | 9 - 10 | 2234 | 78 | ||
| XM_022411051.1 | XP_022266759.1 | 7 - 8 | 1091 | 78 | ||
| XM_022411052.1 | XP_022266760.1 | 7 - 8 | 931 | 78 | ||
| XM_534832.6 | XP_534832.4 | 8 - 9 | 2015 | 78 | ||
More Information
Set Membership: |
|

PUS7L
Genomic Map